missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMIGD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA538057
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
A recommended positive control is rat liver tissue lysate. At least two isoforms of TMIGD1 are known to exist; this antibody will detect both isoforms.
TMIGD1 (transmembrane and immunoglobulin domain containing 1) is a 262 amino acid single-pass membrane protein that contains two Ig-like C2-type (immunoglobulin-like) domains. It is a nonsecreted, glycosylated protein located in the cytoplasm and cell membrane. TMIGD1 might be a possible cell-differentiation marker in the intestine, whose expression is lost during cellular transformation and promote progression of tumorigenesis in the intestinal epithelium. TMIGD1 has also been postulated to play a role in cell adhesion.
Spécification
| TMIGD1 | |
| Polyclonal | |
| Unconjugated | |
| TMIGD1 | |
| 2010002A20Rik; AWKS9372; RGD1307444; TMIGD; Tmigd1; transmembrane and immunoglobulin domain containing 1; transmembrane and immunoglobulin domain-containing protein 1; UNQ9372; UNQ9372/PRO34164 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 363654, 388364, 66601 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q6UXZ0, Q9D7L8 | |
| TMIGD1 | |
| 18 amino acid peptide near the center of human TMIGD1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu