missing translation for 'onlineSavingsMsg'
Learn More

TNFAIP8 Antibody (01), PE, Novus Biologicals™

Code produit 18676987 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.1 mL
Conditionnement:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18676987 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18676987 Fournisseur Novus Biologicals Code fournisseur NBP306584PE

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Mouse Monoclonal Antibody

TNFAIP8 Monoclonal antibody specifically detects TNFAIP8 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigène TNFAIP8
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 01
Conjugué PE
Dilution Immunohistochemistry-Paraffin
Formule PBS
Alias de gène GG2-1, Head and neck tumor and metastasis-related protein, MDC-3.13TNF-induced protein GG2-1, NDED, NF-kappa-B-inducible DED-containing protein, SCC-S2SCCS2, TNF alpha-induced protein 8, tumor necrosis factor alpha-induced protein 8, tumor necrosis factor, alpha-induced protein 8
Espèces hôtes Mouse
Immunogène This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TNFAIP8 (Uniprot#: O95379; Met1-Ile198).
Méthode de purification Protein A purified
Quantité 0.1 mL
État réglementaire RUO
Disciplines de recherche Apoptosis
Primaire ou secondaire Primary
Identification génétique (Entrez) 25816
Espèces cibles Human
Contenu et stockage Store at 4C in the dark.
Forme Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.