missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM20 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
Marque: Novus Biologicals NBP1-81556
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TOMM20 Polyclonal specifically detects TOMM20 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spécification
| TOMM20 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Simple Western, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 0.25-2 μg/mL, SDS-Page 1:100 - 1:2000 | |
| KIAA0016Outer mitochondrial membrane receptor Tom20, MAS20, MGC117367, Mitochondrial 20 kDa outer membrane protein, mitochondrial import receptor subunit TOM20 homolog, MOM19, TOM20, translocase of outer mitochondrial membrane 20 homolog (yeast), translocase of outer mitochondrial membrane 20 homolog type II | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9804 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TOMM20 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KRRSDPNFKNRLRERRKKQKLAKERAGLSKLPDLKDAEAVQKFFLEEIQLGEELLAQGEYEKGVDHLTNAIAVCGQPQQLLQVLQQTLPPPVFQMLLTKLPTISQRIVSAQSLAEDDV | |
| 0.1 mL | |
| Primary | |
| Specificity of human TOMM20 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu