Learn More
Invitrogen™ TRAIL-R2 (DR5) Monoclonal Antibody (K70030_15E10), Invitrogen™
Mouse Monoclonal Antibody
438.00€ - 658.00€
Spécification
| Antigène | TRAIL-R2 (DR5) |
|---|---|
| Clone | K70030_15E10 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
30193933
|
Invitrogen™
MA555733 |
50 μg |
438.00€
50µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
30194648
|
Invitrogen™
MA556113 |
200 μg |
658.00€
200µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Sequence of this protein is as follows: SGEVELSPCT TTRNTVCQCE EGTFREEDSP EMCRKCRTGC PRGMVKVGDC TPWSDIECVH KESGTKHSGE
TRAIL-R2 (DR5) Monoclonal antibody specifically detects TRAIL-R2 (DR5) in Human samples. It is validated for Western BlotSpécification
| TRAIL-R2 (DR5) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide | |
| apoptosis inducing protein TRICK2A/2B; apoptosis inducing receptor TRAIL-R2; CD262; cytotoxic TRAIL receptor-2; death domain containing receptor for TRAIL/Apo-2L; death receptor 5; Dr5; dr-5; Fas-like protein; KILLER; KILLER/DR5; KILLER/DR5 TRAIL death-inducing receptor; Ly98; MK; p53-regulated DNA damage-inducible cell death receptor(killer); sCD262; soluble CD262; TNF receptor superfamily member 10b; TNF-related apoptosis-inducing ligand receptor 2; TNFRSF10B; TRAIL receptor 2; TRAILR2; TRAIL-R2; TRICK2; TRICK2A; TRICK2B; TRICKB; tumor necrosis factor receptor superfamily member 10B; tumor necrosis factor receptor superfamily, member 10b; tumor necrosis factor receptor-like protein ZTNFR9; UNQ160/PRO186; ZTNFR9 | |
| Recombinant human TNFRSF10B (immunogen range is 121-190). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| TNFRSF10B |
| K70030_15E10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O14763 | |
| 8795 | |
| IgG2b | |
| Protein A | |
| Antibody |