Learn More
TREM1 Antibody (L5-B8.2A12.3A12), Allophycocyanin, Novus Biologicals™
Rat Monoclonal Antibody
Marque:  Novus Biologicals NBP1-40907APC
Code nomenclature Nacres: NA.46
Code produit. 18246792
  • 589.00€ / 0.25mL
Description
TREM1 Monoclonal specifically detects TREM1 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
TREM1
Monoclonal
APC
CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
Rat
Protein G purified
RUO
Primary
Mouse
Purified
Flow Cytometry
L5-B8.2A12.3A12
Flow Cytometry
TREM1
Mouse TREM-1 Fc fusion protein.
0.25 mL
Signal Transduction
54210
Store at 4C in the dark.
IgG2a
For Research Use Only