Learn More
Abnova™ TRIM25 (Human) Recombinant Protein
Human TRIM25 partial ORF ( NP_005073, 211 a.a. - 310 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00007706-Q01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
- Sequence: SQINGASRALDDVRNRQQDVRMTANRKVEQLQQEYTEMKALLDASETTSTRKIKEEEKRVNSKFDTIYQILLKKKSEIQTLKEEIEQSLTKRDEFEFLEK
Spécification
NP_005073 | |
tripartite motif-containing 25 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TRIM25 | |
GST |
7706 | |
Wheat germ expression system | |
10 μg | |
EFP, RNF147, Z147, ZNF147 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |