missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TROP-2 Antibody (Sacituzumab-MMAE) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Marque: Novus Biologicals™ NBP3-28116-1mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TROP-2 Monoclonal antibody specifically detects TROP-2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
| TROP-2 | |
| Monoclonal | |
| Unconjugated | |
| P09758 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Sacituzumab-MMAE | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, TROP2GA7331, tumor-associated calcium signal transducer 2 | |
| TROP2 | |
| 1 mg | |
| Signal Transduction | |
| 4070 | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu