missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROP-2 Antibody (TACSTD2/2152), Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-75745-20ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TROP-2 Monoclonal antibody specifically detects TROP-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Spécification
| TROP-2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 μg/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| P09758, P09758, P09758 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array | |
| TACSTD2/2152 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, TROP2GA7331, tumor-associated calcium signal transducer 2 | |
| Recombinant fragment of human TROP-2 protein (around aa 31-274) (exact sequence is proprietary) (Uniprot: P09758 ) | |
| 20 μg | |
| Signal Transduction | |
| 4070 | |
| Store at 4°C. | |
| IgG1 κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu