missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROP-2 Antibody (TACSTD2/7348R), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP3-20920AF488
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TROP-2 Monoclonal antibody specifically detects TROP-2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spécification
| TROP-2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, TROP2GA7331, tumor-associated calcium signal transducer 2 | |
| Recombinant fragment of human TROP-2 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Signal Transduction | |
| 4070 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| TACSTD2/7348R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction