Description
TRP2 Polyclonal antibody specifically detects TRP2 in Human, Rat samples. It is validated for Western Blot
Spécification
Spécification
| Antigène | TRP2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Dilution | Western Blot 1:500 - 1:1000 |
| Formule | PBS (pH 7.3), 50% glycerol |
| Alias de gène | dopachrome delta-isomerase, dopachrome tautomerase (dopachrome delta-isomerase, tyrosine-related protein 2), DT, EC 5.3.3.12, L-dopachrome tautomerase, TRP2, TRP-2, Tyrosinase-related protein 2, TYRP2, TYRP2L-dopachrome Delta-isomerase |
| Espèces hôtes | Rabbit |
| Immunogène | Recombinant fusion protein containing a sequence corresponding to amino acids 50-350 of human DCT (NP_001913.2). NVCGSQQGRGQCTEVRADTRPWSGPYILRNQDDRELWPRKFFHRTCKCTGNFAGYNCGDCKFGWTGPNCERKKPPVIRQNIHSLSPQEREQFLGALDLAKKRVHPDYVITTQHWLGLLGPNGTQPQFANCSVYDFFVWLHYYSVRDTLLGPGRPYRAIDFSHQGPAFVTWHRYHLLCLERDLQRLIGNESFALPYWNFATGRNECDVCTDQLFGAARPDDPTLISRNSRFSSWETVCDSLDDYNHLVTLCNGTYEGLLRRNQMGRNSMKLPTLKDIRDCLSLQKFDNPPFFQNSTFSFRNA |
| Méthode de purification | Affinity purified |
| Afficher plus |