missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tryptase Monoclonal Antibody (C2), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen™ MA551170
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Tryptase is a protein that belongs to the family of serine proteases and is primarily found in mast cells. It plays a crucial role in the immune response by cleaving and activating other proteins involved in inflammation and tissue repair. Tryptase is also involved in the regulation of blood pressure and has been implicated in various diseases, including asthma, allergies, and cardiovascular disorders. Its structure consists of four identical subunits that form a tetramer, and it is activated by proteolytic cleavage of its proenzyme form.
Spécification
| Tryptase | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q15661, Q9N2D1 | |
| MCT7, Tpsab1 | |
| Recombinant Human TPS protein, Gly161-Tyr262 (Accession #Q15661-1). | |
| 20 μL | |
| Primary | |
| Canine, Human, Pig | |
| Antibody | |
| IgG2a κ |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| C2 | |
| Unconjugated | |
| Tpsab1 | |
| TPSG1/TPSB2/TPSAB1/TPSD1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 397389, 7177 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu