missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tryptase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
185.00€ - 393.00€
Spécification
| Antigène | Tryptase |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
17980663
|
Invitrogen™
PA5145362 |
20 μL |
185.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
17980673
|
Invitrogen™
PA5145361 |
200 μL |
393.00€
200µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Tryptase is a protein that belongs to the family of serine proteases and is primarily found in mast cells. It plays a crucial role in the immune response by cleaving and activating other proteins involved in inflammation and tissue repair. Tryptase is also involved in the regulation of blood pressure and has been implicated in various diseases, including asthma, allergies, and cardiovascular disorders. Its structure consists of four identical subunits that form a tetramer, and it is activated by proteolytic cleavage of its proenzyme form.Spécification
| Tryptase | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| P27435, Q02844 | |
| 100503895, 54271 | |
| Recombinant Mouse TPS protein, Ser149-Phe273 (Accession #Q02844). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| TPSG1/TPSB2/TPSAB1/TPSD1 | |
| Tpsab1 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Tpsab1 |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit