Learn More
TSC1 Antibody (488915), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals FAB4379J
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TSC1 Monoclonal antibody specifically detects TSC1 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Spécification
| TSC1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 488915 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| KIAA0243MGC86987, LAMhamartin, TSC, tuberous sclerosis 1, Tuberous sclerosis 1 protein, tumor suppressor | |
| E. coli-derived recombinant human TSC1, Asp156-Thr300, Accession # Q92574 | |
| 0.1 mL | |
| Autophagy, Cancer, Hypoxia, mTOR Pathway | |
| 7248 | |
| Store at 4°C in the dark. | |
| IgG2b |