Learn More
Invitrogen™ TSH beta Monoclonal Antibody (155)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA182909
Description
MA1-82909 is specific for beta (TSH) and shows minimal cross-reactivity with beta Human chorionic gonadotropin (hCG), Luteinizing Hormone (LH) or Follicle Stimulating Hormone (FSH). Mouse anti Human thyroid stimulating hormone, clone 155 recognizes the beta chain of human beta (TSH) and shows minimal cross-reactivity with human chorionic gonadotropin (hCG), luteinizing hormone (hLH) or follicle stimulating hormone (hFSH).
TSH is a pituitary hormone of 28kDa, which stimulates thyroid growth and production of thyroid hormones. This antibody labels thyrotropic cells of the pituitary.
Spécification
| TSH beta | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with <0.1% sodium azide; pH 7.4 | |
| P01222 | |
| TSHB | |
| Native human beta-TSH. | |
| 1 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 155 | |
| Unconjugated | |
| TSHB | |
| thyroid stimulating hormone beta; thyroid stimulating hormone subunit beta; thyroid stimulating hormone, beta; thyroid stimulating hormone, beta subunit; thyroid-stimulating hormone beta subunit; thyroid-stimulating hormone subunit beta; Thyrotropin alfa; Thyrotropin beta chain; thyrotropin beta subunit; thyrotropin subunit beta; TSH; Tshb; TSH-B; TSH-beta | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7252 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.