missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTF1 Mouse anti-Human, Porcine, Clone: 2C8F3, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66034-1-IG-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a protein initially identified as a thyroid-specific transcription factor. The encoded protein binds to thyroglobulin promoter and regulates the expression of thyroid-specific genes but has also been shown to regulate the expression of genes involved in morphogenesis. Mutations and deletions in this gene are associated with benign hereditary chorea, choreoathetosis, congenital hypothyroidism, and neonatal respiratory distress, and may be associated with thyroid cancer. Multiple transcript variants encoding different isoforms have been found for this gene.Spécification
| TTF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P43699 | |
| NKX2-1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG3 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2C8F3 | |
| Unconjugated | |
| NKX2-1 | |
| BHC, Homeobox protein Nkx 2.1, NK 2, NK2 homeobox 1, NKX2 1, NKX2.1, NKX2-1, NKX2A, TEBP, Thyroid nuclear factor 1, Thyroid transcription factor 1, TITF1, TTF 1, TTF1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 100154384, 7080 | |
| -20°C | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu