Learn More
Tyrosine Hydroxylase Antibody, Novus Biologicals™
Sheep Polyclonal Antibody has been used in 77 publications
Marque:  Novus Biologicals NB300-110
Code nomenclature Nacres: NA.46
Code produit. 18740954
  • 622.00€ / 0.10mL
Description
Tyrosine Hydroxylase Polyclonal specifically detects Tyrosine Hydroxylase in Human, Mouse, Rat, Amphibian, Avian, Mammal, Rabbit, Reptile samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, Single Cell Western.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
TH
60 kDa
Human, Mouse, Rat