Learn More
Tyrosine Hydroxylase Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Marque: Novus Biologicals NBP3-05555-50ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Tyrosine Hydroxylase Polyclonal antibody specifically detects Tyrosine Hydroxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Spécification
| Tyrosine Hydroxylase | |
| Polyclonal | |
| Western Blot 1:50000, Immunohistochemistry 1:5000, Immunocytochemistry/ Immunofluorescence 1:5000 | |
| DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase | |
| Full length human Tyrosine Hydroxylase as expressed in and purified from E. coli. | |
| 50 μg | |
| Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Prostate Cancer, Tyrosine Kinases | |
| 7054 | |
| Store at 4°C. | |
| IgY |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Supplied as an aliquot of IgY preparation | |
| Chicken | |
| IgY purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |