Learn More
Ubiquitin Antibody (Ubi-1), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NB300-130PCP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Ubiquitin Monoclonal antibody specifically detects Ubiquitin in Human, Mouse, Rat, Porcine, Bovine, C. elegans, Chicken, Drosophila, Equine, Plant, Monkey, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Spécification
| Western Blot | |
| Purified bovine erythrocyte ubiquitin cross-linked with glutaraldehyde to KLH | |
| RUO | |
| Pig |
| PerCP | |
| Affinity purified | |
| 6233 |