Learn More
UNC13D/Munc13-4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16905-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a protein that is a member of the UNC13 family, containing similar domain structure as other family members but lacking an N-terminal phorbol ester-binding C1 domain present in other Munc13 proteins. The protein appears to play a role in vesicle maturation during exocytosis and is involved in regulation of cytolytic granules secretion. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis type 3, a genetically heterogeneous, rare autosomal recessive disorder.Spécification
| UNC13D/Munc13-4 | |
| Polyclonal | |
| Unconjugated | |
| UNC13D | |
| FHL3, HLH3, HPLH3, Munc13 4, Munc13-4, Protein unc 13 homolog D, unc 13 homolog D (C. elegans), UNC13D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 201294 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q70J99 | |
| UNC13D | |
| UNC13D/Munc13-4 Fusion Protein Ag10519 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |