Learn More
Invitrogen™ Utomilumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA558413
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Utomilumab is under investigation in clinical trial NCT03318900 (T Cell Immunotherapy for Advanced Ovarian Cancer).
Spécification
| Utomilumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PF-2566; PF-05082566 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG2SA |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |