Learn More
Vβ 10[b] TCR Rat anti-Mouse, BUV395, Clone: B21.5, BD Optibuild™
Rat Monoclonal Antibody
Marque: BD Biosciences 742825
Description
The B21.5 monoclonal antibody specifically recognizes the Vβ10 T-Cell Receptor (TCR) of mice having the b /haplotype (e.g., A, AKR, BALB/c, CBA, C3H/He, C57BL, C58, DBA/1, DBA/2) of the Tcrb /gene complex. It does not react with mice having the a /(eg, C57BR, C57L, SJL, SWR) or c /(eg, RIII) Tcrb /haplotype.
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
Spécification
| Vβ 10[b] TCR | |
| B21.5 | |
| Brilliant Ultraviolet 395 | |
| TCR Vβ10, TCR V beta 10, TCR Vb10 | |
| Mouse Cytotoxic T-Cell Clone Soluble αβ TCR | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2a λ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.