Learn More
Vβ 13 TCR Mouse anti-Mouse, Brilliant Violet 510, Clone: MR12-3, BD Optibuild™
Mouse Monoclonal Antibody
Marque: BD Biosciences 743991
Description
The MR12-3 monoclonal antibody specifically binds to the Vβ13 T-cell Receptor (TCR) of mice having the b /haplotype (e.g., A, AKR, BALB/c, CBA, C3H/He, C57BL, C58, DBA/1, DBA/2) of the Tcrb /gene complex. The Tcrb-V13 /gene locus is deleted in mice having the a /(e.g., C57BR, C57L, SJL, SWR) or c /(e.g., RIII) haplotype.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
Spécification
| Vβ 13 TCR | |
| MR12-3 | |
| Brilliant Violet 510 | |
| TCR V beta 13, TCR Vβ 13, TCR Vβ13, Vβ 13 T-cell Receptor | |
| Not reported | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG1 κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.