Learn More
Novus Biologicals™ Varicella Zoster Virus Glycoprotein E Antibody (09) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals™ NBP3-26914-20ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Varicella Zoster Virus Glycoprotein E Monoclonal antibody specifically detects Varicella Zoster Virus Glycoprotein E in Virus samples. It is validated for ELISA,Western Blot,Sandwich ELISA Detection
Spécification
| Varicella Zoster Virus Glycoprotein E | |
| Monoclonal | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Western Blot, Sandwich ELISA Detection | |
| 09 | |
| Western Blot 0.2-1 μg/mL, ELISA 0.5-1 μg/mL, Sandwich ELISA Detection 0.1-1 μg/mL | |
| Envelope glycoprotein E, GE, ORF68 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Varicella Zoster Virus Glycoprotein E (Uniprot#: Q9J3M8; Met1-Tyr538). | |
| 20 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |