Learn More
Varicella Zoster Virus Glycoprotein E Antibody (144), Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP3-31977-20ug
166.60 EUR valable jusqu'au 2025-12-16
MEILLEUR PRIX promo! Utilisez le code promo "24090" pour bénéficier de cette offre.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Varicella Zoster Virus Glycoprotein E Monoclonal antibody specifically detects Varicella Zoster Virus Glycoprotein E in Virus samples. It is validated for ELISA,Western Blot
Spécification
| Varicella Zoster Virus Glycoprotein E | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Western Blot | |
| 144 | |
| Western Blot 0.1-1 μg/mL, ELISA 0.1-0.2 μg/mL | |
| Envelope glycoprotein E, GE, ORF68 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Varicella Zoster Virus Glycoprotein E (Uniprot#: Q9J3M8; Met1-Tyr538) | |
| 20 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |