Learn More
PeproTech VEGF-165 Polyclonal Antibody, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
219.00€ - 1834.00€
Spécification
| Antigène | VEGF-165 |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
17884733
![]() |
PeproTech
500-P131-50UG |
50 μg |
219.00€
50µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
17894143
|
PeproTech
500-P131-100UG |
100 μg |
275.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
17863953
|
PeproTech
500-P131-1MG |
1 mg |
1834.00€
1mg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
AA Sequence of recombinant protein: MAPTTEGEQK SHEVIKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNITMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPEKHCEPC SERRKHLFVQ DPQTCKCSCK NTDSRCKARQ LELNERTCRC DKPRR. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine VEGF 165. Anti-Murine VEGF 165-specific antibody was purified by affinity chromatography employing an immobilized Murine VEGF 165 matrix. Sandwich ELISA: To detect Murine VEGF 165 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Murine VEGF 165 (500-P131Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine VEGF 165. Western Blot: To detect Murine VEGF 165 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Murine VEGF165 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
The vascular endothelial growth factor (VEGF) family currently includes VEGF-A, VEGF-B, VEGF-C, VEGF-D, VEGF-E, and PIGF. VEGF and its receptor system have been shown to be the fundamental regulators in the cell signaling of angiogenesis. Most tumors have the absolute requirement of angiogenesis and VEGF has been described as the most potent angiogenic cytokine linked to this process. To date 5 different isoforms of VEGF have been described. These isoforms are generated as the result of alternative splicing from a single VEGF gene. These various isoforms have been shown to bind to two tyrosine-kinase receptors flt-1 (VEGFR-1) and flk-1/KDR (VEGFR-2), which have been found to be expressed almost exclusively on endothelial cells.Spécification
| VEGF-165 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Q00731 | |
| Vegfa | |
| Primary | |
| -20°C |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| 22339 | |
| E.coli-derived Recombinant Murine VEGF 165. | |
| Antigen affinity chromatography | |
| Vegfa |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.