Learn More
CD31/PECAM-1 Antibody (LCI-4), Alexa Fluor™ 488, Novus Biologicals™
Tous les produits Bio Techne ProduitsDescription
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Spécification
Spécification
| Antigène | CD31/PECAM-1 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | LCI-4 |
| Conjugué | Alexa Fluor 488 |
| Dilution | Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen |
| Formule | 50mM Sodium Borate with 0.05% Sodium Azide |
| Alias de gène | adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA′, PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule |
| Symboles de gène(s) | PECAM1 |
| Espèces hôtes | Mouse |
| Afficher plus |
For Research Use Only