Learn More
CXCL5/ENA-78 Antibody (32B3), PE, Novus Biologicals™
Rat Monoclonal Antibody
Marque: Novus Biologicals NBP2-22026PE
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CXCL5/ENA-78 Monoclonal antibody specifically detects CXCL5/ENA-78 in Mouse samples. It is validated for Western Blot, ELISA, Neutralization
Spécification
| CXCL5/ENA-78 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, ELISA, Neutralization | |
| 32B3 | |
| Western Blot, ELISA, Block/Neutralize | |
| chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78 | |
| Mouse recombinant CXCL5/ENA-78 | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines | |
| 6374 | |
| Store at 4C in the dark. | |
| IgG2 |