Learn More
SARM1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-29625C
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SARM1 Polyclonal antibody specifically detects SARM1 in Human samples. It is validated for Western Blot, Immunofluorescence
Spécification
| SARM1 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| KIAA0524SAM domain-containing protein 2, SAMD2sterile alpha and HEAT/Armadillo motif protein, ortholog of Drosophila, SARMFLJ36296, Sterile alpha and Armadillo repeat protein, sterile alpha and TIR motif containing 1, sterile alpha and TIR motif-containing protein 1, Sterile alpha motif domain-containing protein 2, Tir-1 homolog | |
| Synthetic peptide made to an internal portion of the human SARM protein (between residues 300-350) [NCBI NP_055892] | |
| 0.1 mL | |
| Innate Immunity | |
| 23098 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |