Learn More
HIV-1 gp120 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP3-06630MFV500
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
HIV-1 gp120 Polyclonal antibody specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISA
Spécification
| HIV-1 gp120 | |
| Polyclonal | |
| Western Blot, ELISA | |
| Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein | |
| Produced in rabbits immunized with purified, recombinant HIV-1 gp120 (Ala29-Glu502; Accession#:AFM77980.1). | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| 155971 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |