Description
MOX1 Polyclonal antibody specifically detects MOX1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Spécification
Spécification
| Antigène | MOX1 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugué | PerCP |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence |
| Formule | PBS |
| Alias de gène | homeobox protein MOX-1, mesenchyme homeobox 1MOX1mesenchyme homeo box 1 |
| Espèces hôtes | Rabbit |
| Immunogène | Antibody was raised against a 19 amino acid synthetic peptide near the center of human MOX1. The immunogen is located within amino acids 140 - 190 of MOX1. Amino Acid Squence: SRRRKESSDNQENRGKPEG |
| Méthode de purification | Peptide affinity purified |
| Afficher plus |