Description
Ractopamine Monoclonal specifically detects Ractopamine in Non-species specific samples. It is validated for ELISA.
Spécification
Spécification
| Antigène | Ractopamine |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | Rac1 |
| Conjugué | Unconjugated |
| Dilution | ELISA 1:10000 |
| Formule | Ascites with 0.03% Sodium Azide |
| Alias de gène | C18H23NO3 |
| Espèces hôtes | Mouse |
| Immunogène | Rac-OVA. |
| Afficher plus |
For Research Use Only