Learn More
IL-6 Antibody (1936), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals FAB2061MFV500
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IL-6 Monoclonal antibody specifically detects IL-6 in Human samples. It is validated for Immunohistochemistry, Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF
Spécification
| IL-6 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Adaptive Immunity, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Innate Immunity, Neuroscience | |
| 3569 | |
| Store at 4C in the dark. |
| Immunohistochemistry, Neutralization, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 1936 | |
| Immunohistochemistry, Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6 | |
| E. coli-derived recombinant human IL-6, Val30-Met212, Accession # P05231 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |