Description
EIF4E3 Polyclonal specifically detects EIF4E3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Spécification
Spécification
| Antigène | EIF4E3 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunofluorescence |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL |
| Formule | PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide |
| Alias de gène | eIF4E type 3, eIF-4E type 3, eIF-4E3, eIF4E-3, eukaryotic translation initiation factor 4E family member 3, eukaryotic translation initiation factor 4E member 3, eukaryotic translation initiation factor 4E type 3, MGC39820, MGC86971 |
| Symboles de gène(s) | EIF4E3 |
| Espèces hôtes | Rabbit |
| Immunogène | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LLATIGEQFTDCAAADDEVIGVSVSVRDREDVVQVWNVNASLVGEATVLEKIY |
| Afficher plus |
For Research Use Only