Learn More
TIAM1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NB100-2301V
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TIAM1 Polyclonal antibody specifically detects TIAM1 in Human, Mouse samples. It is validated for Western Blot
Spécification
| TIAM1 | |
| Polyclonal | |
| Western Blot | |
| FLJ36302, human T-lymphoma invasion and metastasis inducing TIAM1 protein, 10TIAM-1, T-cell lymphoma invasion and metastasis 1, T-lymphoma invasion and metastasis-inducing protein 1 | |
| The immunogen recognized by this antibody maps to a region between residues 1550 and the C-terminus (residue 1591) of human T-cell lymphoma invasion and metastasis 1 using the numbering given in entry NP_003244.1 (GeneID 7074) | |
| 0.1 mL | |
| Cancer | |
| 7074 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |