Learn More
LGR5 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 30007-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The LGR5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LGR5. This antibody recognizes human, mouse, rat antigen. The LGR5 antibody has been validated for the following applications: WB, ELISA analysis.Spécification
| LGR5 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8549 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, ELISA | |
| 750 μg/mL | |
| FEX; GPR49; GPR67; GRP49; HG38; MGC117008 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 100 kDa | |
| Liquid |