missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Antibody (CL0157) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-52867JF525
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Gene Knock-Out
Spécification
| Vimentin | |
| Monoclonal | |
| Janelia Fluor 525 | |
| epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin | |
| This antibody was developed using a recombinant protein derived from P08670, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
| 7431 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Gene Knock-Out | |
| CL0157 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu