Learn More
Vimentin Antibody, Novus Biologicals™
Chicken Polyclonal Antibody has been used in 44 publications
Marque:  Novus Biologicals NB300-223
Code nomenclature Nacres: NA.46
Code produit. 18738703
  • 567.00€ / 0.10mL
Description
Vimentin Polyclonal specifically detects Vimentin in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Polyclonal
FLJ36605, vimentin
Precipitation
Western Blot 1:10000-1:20000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:5000, Immunohistochemistry-Paraffin 1:150, Immunohistochemistry-Frozen
Human, Mouse, Rat, Pig, Bovine
IgY
For Research Use Only