missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VISTA Mouse anti-Human, BV510, Clone: MIH65.rMAb (also known as MIH65), BD Biosciences™
Mouse Monoclonal Antibody
Marque: BD Biosciences 749654
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
VISTA Monoclonal antibody specifically detects VISTA in Human samples. It is validated for Flow Cytometry
Spécification
| VISTA | |
| Monoclonal | |
| Brilliant Violet 510 | |
| VSIR; B7-H5; B7H5; C10orf54; DD1alpha; GI24; SISP1 | |
| Human VISTA Transfected Cell Line | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| MIH65.rMAb (also known as MIH65) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV510 under optimal conditions that minimize unconjugated dye and antibody. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu