missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Vitamin B6 Monoclonal Antibody (2F9)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA549648
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Antibody tested in indirect ELISA.
Vitamin B6, also known as pyridoxine, is a water-soluble vitamin that is essential for the proper functioning of the body. It is involved in a variety of metabolic processes, including the production of neurotransmitters, the synthesis of hemoglobin, and the metabolism of amino acids and carbohydrates. Vitamin B6 is found in a variety of foods, including meat, fish, poultry, whole grains, and vegetables such as spinach and potatoes. Vitamin B6 deficiency is rare, but it can lead to a range of symptoms, including anemia, skin inflammation, and nervous system disorders.
Spécification
| Vitamin B6 | |
| Monoclonal | |
| 0.82 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 2F9 | |
| Unconjugated | |
| Pyridoxine | |
| Conjugate of VB5 hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu