Learn More
VLDL-Receptor Mouse, Clone: VLDLR/1337, enQuireBio™
Mouse Monoclonal Antibody
Marque:  EnquireBio 7436-MSM1-P1
Code nomenclature Nacres: NA.46
Code produit. 15981068
  • ONLINE EXCLUSIVE PRICE
    554.40€
    478.00€ / 100µg
    Économisez 76.40€
    13% de réduction
Spécification
VLDL-Receptor
Cytoplasmic
VLDLR/1337
Unconjugated
NP_001309154
Mouse
143-161kDa
Protein A/G Purified
RUO
7436
IgG1
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
VLDLR
Recombinant human VLDLR fragment from c-terminal
HeLa or U-251 cells. Pancreas, Cerebellum, Heart or Skeletal Muscle.
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.