missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Parvovirus VP2 Monoclonal Antibody (4E5A4)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA538659
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody has been tested in indirect ELISA.
Isoform VP2 is a structural protein that resides within the core of the capsid surrounded by 72 VP1 pentamers. Participates in host cell receptor binding together with VP1. Following virus endocytosis and trafficking to the endoplasmic reticulum, VP2 and VP3 form oligomers and integrate into the endoplasmic reticulum membrane. Heterooligomer VP2-VP3 may create a viroporin for transporting the viral genome across the endoplasmic reticulum membrane to the cytoplasm. Nuclear entry of the viral DNA involves the selective exposure and importin recognition of VP2 or Vp3 nuclear localization signal (shared C-terminus). Plays a role in virion assembly within the nucleus in particular through a DNA-binding domain located in the C-terminal region. A N-terminal myristoylation suggests a scaffold function for virion assembly.
Spécification
| Human Parvovirus VP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Purified recombinant fragment of parovirus VP2 (aa: 296-438) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4E5A4 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu