missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS4B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17673-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Class E vacuolar protein sorting (vps) proteins are necessary for appropriate sorting of receptors in the yeast endocytic pathway. The yeast Vps4p is a member of the AAA protein family (ATPases associated with diverse cellular activities) and plays an important role in transporting proteins out of a prevacuolar endosomal compartment. In human, two non-allelic orthologous proteins (VPS4A and VPS4B) of yeast Vps4p are known and can form heteromeric complexes with each other. Both VPS4 (also known as SKD1 in mouse) proteins are class E VPSs and are involved in intracellular protein trafficking, similar to Vps4p in yeast. A human CHMP1 protein, which is implicated in multivesicular body formation, physically interacts with VPS4. HIV-1 uses cellular machinery to bud from infected cells and requires VPS4 and TSG101/VPS23 for this budding process. Domit negative mutant of VPS4 inhibit vacuolar protein sorting and also arrest HIV-1 and MLV budding. Thus, retroviruses normally use the VPS pathway to form multivesicular bodies during the budding process.Spécification
| VPS4B | |
| Polyclonal | |
| Unconjugated | |
| VPS4B | |
| MIG1, Protein SKD1, SKD1, VPS4 2, VPS42, VPS4B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20479, 360834, 9525 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75351, P46467 | |
| Vps4b | |
| VPS4B Fusion Protein Ag11610 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu