missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VSIG4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA552018
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: PPISYIWYKQ QTNNQEPIKV ATLSTLLFKP AVIADSGSYF CTAKGQVGSE QHSDIVKFVV KDSSKLLKTK TEAPTTMTYP LKATSTVKQS WDWTTDMDGY L Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 25%.
T cell activation by APCs is positively and negatively regulated by members of the B7 family. VSIG4 is a strong negative regulator of murine and human T cell proliferation and IL-2 production.
Spécification
| VSIG4 | |
| Polyclonal | |
| Unconjugated | |
| VSIG4 | |
| A530061A11; BC025105; complement receptor of the immunoglobulin superfamily; CRIg; Ig superfamily protein; MGC137243 protein; protein Z39Ig; UNQ317/PRO362; V-set and immunoglobulin domain containing 4; V-set and immunoglobulin domain-containing protein 4; VSIG4; Z39IG | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11326 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y279 | |
| VSIG4 | |
| Recombinant protein corresponding to Human VSIG4. Recombinant protein control fragment (Product #RP-92217). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu