missing translation for 'onlineSavingsMsg'
Learn More

WASF1/WAVE1 Antibody [mFluor Violet 450 SE], Novus Biologicals™

Code produit 30651950 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.1 mL
Conditionnement:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
30651950 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30651950 Supplier Novus Biologicals Supplier No. AF5514MFV450

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Goat Polyclonal Antibody

WASF1/WAVE1 Polyclonal antibody specifically detects WASF1/WAVE1 in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigène WASF1/WAVE1
Applications Western Blot
Classification Polyclonal
Conjugué mFluor Violet 450 SE
Formule 50mM Sodium Borate
Alias de gène KIAA0269WAVEhomology of dictyostelium scar 1, Protein WAVE-1, SCAR1WASP family protein member 1, Verprolin homology domain-containing protein 1, WAS protein family, member 1, WAVE1FLJ31482, wisk
Espèces hôtes Goat
Immunogène E. coli-derived recombinant human WASF1/WAVE1, Met1-Tyr250, Accession # Q92558
Méthode de purification Antigen Affinity-purified
Quantité 0.1 mL
État réglementaire RUO
Disciplines de recherche Phospho Specific
Primaire ou secondaire Primary
Identification génétique (Entrez) 8936
Espèces cibles Human, Mouse
Contenu et stockage Store at 4°C in the dark.
Forme Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.