Learn More
WDR68 Antibody (SR1650), Novus Biologicals™
Code produit 18612025 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
100 μg
50 μg
Conditionnement:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18612025 50 μg 50µL
18692795 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18612025 Fournisseur Novus Biologicals Code fournisseur NBP32249250ul
Rabbit Monoclonal Antibody
WDR68 Monoclonal antibody specifically detects WDR68 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Spécification
Antigène WDR68
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone SR1650
Conjugué Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200
Formule PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Alias de gène DDB1 and CUL4 associated factor 7, DDB1- and CUL4-associated factor 7, HAN11AN11, human anthocyanin, seven-WD-repeat protein of the AN11 family-1, SWAN-1, WD repeat domain 68, WD repeat-containing protein 68, WD repeat-containing protein An11 homolog, WDR68, WD-repeat protein
Espèces hôtes Rabbit
Immunogène A synthesized peptide derived from human WDR68 (Uniprot #: P61962)
Méthode de purification Affinity purified
Quantité 50 μg
État réglementaire RUO
Disciplines de recherche Cell Biology, Signal Transduction
Primaire ou secondaire Primary
Identification génétique (Entrez) 10238
Espèces cibles Human
Contenu et stockage Store at -20° C. Avoid freeze/thaw cycles.
Forme Purified
Isotype IgG