missing translation for 'onlineSavingsMsg'
Learn More
Learn More
West Nile Virus MP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-41052
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
West Nile Virus MP Polyclonal antibody specifically detects West Nile Virus MP in Virus samples. It is validated for ELISA.
Spécification
| West Nile Virus MP | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the middle of the West Nile Virus MP. The immunogen is located within amino acids 190 - 240. Amino Acid Squence: CTKTRHSRRSRRSLT | |
| 0.1 mg | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| ELISA | |
| 1 mg/mL | |
| ELISA 1 μg/mL | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Virus | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu