missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA534320
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Baboon, Monkey (100%) Marmoset, Mouse, Hamster, Guinea pig, Platypus (87%).
WNT2 is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.
Spécification
| WNT2 | |
| Polyclonal | |
| Unconjugated | |
| Wnt2 | |
| 2610510E18Rik; etID309852.18; INT1L 1; Int1l1; int-1-like protein 1; INT-1-related protein; IRP; IRP protein; Mirp; protein Wnt-2; secreted growth factor; wingless-related MMTV integration site 2; wingless-type MMTV integration site 2; wingless-type MMTV integration site family member 2; wingless-type MMTV integration site family, member 2; Wnt; WNT 2; Wnt family member 2; Wnt2; Wnt-2; Wnt2a; ZfWnt2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7472 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P09544 | |
| Wnt2 | |
| Synthetic 15 amino acid peptide from internal region of human WNT2. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu