missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XBP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Marque: Novus Biologicals NBP2-20917
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
XBP1 Polyclonal specifically detects XBP1 in Human, Mouse, Porcine samples. It is validated for Western Blot, Knockout Validated, Knockdown Validated.
Spécification
| XBP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP2Tax-responsive element-binding protein 5 | |
| Rabbit | |
| 29 kDa | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Unfolded Protein Response | |
| 7494 | |
| Human, Mouse, Pig | |
| IgG |
| Western Blot, Immunoassay, KnockDown | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| P17861 | |
| XBP1 | |
| Recombinant protein encompassing a sequence within the center region of human XBP1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: cow (89%), Mouse (82%), Rat (86%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu