missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YB1 Antibody (YBX1/2430), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals (Bio-Techne) NBP3-08774CL1
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
YB1 Monoclonal antibody specifically detects YB1 in Human samples. It is validated for Immunohistochemistry (Paraffin),Protein Array
Spécification
| YB1 | |
| Monoclonal | |
| CoraFluor 1 | |
| BP-8, CBF-A, class II, Y box-binding protein I, CSDA2, CSDB, DBPB CCAAT-binding transcription factor I subunit A, EFI-A, Enhancer factor I subunit A, MDR-NF1, MGC104858, MGC110976, MGC117250, NSEP1 Y-box-binding protein 1, nuclease-sensitive element-binding protein 1, Y box binding protein 1, YB1 DNA-binding protein B, YB-1 Y-box transcription factor, YBX1 | |
| Recombinant human full-length YB1 protein (Uniprot: P67809) | |
| 0.1 mL | |
| Breast Cancer, Cancer, DNA Repair, Epigenetics, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Transcription Factors and Regulators, Translation Control | |
| 4904 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Protein Array | |
| YBX1/2430 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu