missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZDHHC19 Polyclonal specifically detects ZDHHC19 in Human samples. It is validated for Western Blot.
Spécification
Spécification
| Antigène | ZDHHC19 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugué | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formule | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Numéro d’ordre du gène | Q8WVZ1 |
| Alias de gène | DHHC domain containing 19, DHHC19, EC 2.3.1.-, zinc finger, DHHC-type containing 19 |
| Symboles de gène(s) | ZDHHC19 |
| Afficher plus |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?